CH4407083228 ChemBridge Corporation

(2,2-difluoro-1,3-benzodioxol-4-yl)boronic acid

  • MDL number MFCD08062343
  • Cas Number : 126120-87-4
  • Molecular Weight : 201.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4030891

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4030891
Availability details for
Quantity
  • Availability
  • Add to Cart