CH4407083236 ChemBridge Corporation

2,3-piperidinedione

  • MDL number MFCD05670115
  • Cas Number : 41907-06-6
  • Molecular Weight : 113.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4038506

Safety Information

MSDS https://www.hit2lead.com/sds/4038506_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart