CH4407083240 ChemBridge Corporation

4-bromo-2,5-dimethyl-1,3-thiazole

  • MDL number MFCD20491436
  • Cas Number : 90672-80-3
  • Molecular Weight : 192.0768

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4040817

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4040817
Availability details for
Quantity
  • Availability
  • Add to Cart