CH4407083244 ChemBridge Corporation

1-isopropyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD06805218
  • Cas Number : 942631-65-4
  • Molecular Weight : 154.1665

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045275

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4045275
Availability details for
Quantity
  • Availability
  • Add to Cart