CH4407083246 ChemBridge Corporation

4-methyl-5,6,7,8-tetrahydro-2-quinazolinol

  • MDL number MFCD01444097
  • Cas Number : 88267-97-4
  • Molecular Weight : 164.2044

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045968

Safety Information

MSDS https://www.hit2lead.com/sds/4045968_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart