CH4407083253 ChemBridge Corporation

5-chloro-2-methyl-4-pyrimidinol

  • MDL number MFCD00223702
  • Cas Number : 26732-04-7
  • Molecular Weight : 144.559

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4047922

Safety Information

MSDS https://www.hit2lead.com/sds/4047922_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart