CH4407083275 ChemBridge Corporation

rac-(4aS,7aS)-4-methylhexahydropyrrolo[3,4-b][1,4]oxazin-3(2H)-one

  • MDL number MFCD26131687
  • Cas Number : 1932083-00-5
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4058083

Safety Information

MSDS https://www.hit2lead.com/sds/4058083_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart