CH4407083287 ChemBridge Corporation

rac-(4aS,7aR)-4-methylhexahydropyrrolo[3,4-b][1,4]oxazin-3(2H)-one

  • MDL number MFCD26131686
  • Cas Number : 1820570-46-4
  • Molecular Weight : 156.1824

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4071900

Safety Information

MSDS https://www.hit2lead.com/sds/4071900_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart