CH4407083304 ChemBridge Corporation

trans-4-bromotetrahydro-3-thiopheneol 1,1-dioxide

  • MDL number MFCD08706466
  • Cas Number : 20688-40-8
  • Molecular Weight : 215.0656

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4085308

Safety Information

MSDS https://www.hit2lead.com/sds/4085308_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart