CH4407083309 ChemBridge Corporation

1-methyl-2-oxo-1,2-dihydro-3-pyridinecarbaldehyde

  • MDL number MFCD22071244
  • Cas Number : 79138-28-6
  • Molecular Weight : 137.136

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4003630

Safety Information

MSDS https://www.hit2lead.com/sds/4003630_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart