CH4407083324 ChemBridge Corporation

1-methyl-1H-benzimidazole-2-carboxylic acid

  • MDL number MFCD01836869
  • Cas Number : 20572-01-4
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400229

Safety Information

MSDS https://www.hit2lead.com/sds/4400229_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart