CH4407083327 ChemBridge Corporation

1-isopropyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD03130043
  • Cas Number : 920006-32-2
  • Molecular Weight : 154.1665

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7117667

Safety Information

MSDS https://www.hit2lead.com/building-blocks/7117667
Availability details for
Quantity
  • Availability
  • Add to Cart