CH4411837375 ChemBridge Corporation

5-ethyl-1,2,4-oxadiazol-3-amine

  • MDL number MFCD11847810
  • Cas Number : 171006-96-5
  • Molecular Weight : 113.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014409

Safety Information

MSDS https://www.hit2lead.com/sds/4014409_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart