CH4415172729 ChemBridge Corporation

[3-(1H-imidazol-5-yl)phenyl]amine dihydrochloride hydrate

  • MDL number MFCD27966289
  • Cas Number : 51746-93-1
  • Molecular Weight : 250.125

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087745

Safety Information

MSDS https://www.hit2lead.com/sds/4087745_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart