CH4415172730 ChemBridge Corporation

dibenzo[b,d]thien-2-ylboronic acid hydrate

  • MDL number MFCD27966290
  • Cas Number : 668983-97-9
  • Molecular Weight : 246.0899

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2O
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087750

Safety Information

MSDS https://www.hit2lead.com/sds/4087750_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart