CH4415172736 ChemBridge Corporation

{2-[3-(2-methylphenyl)-1-pyrrolidinyl]ethyl}amine dihydrochloride

  • MDL number MFCD27966297
  • Cas Number : 1559059-73-2
  • Molecular Weight : 277.2332

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087770

Safety Information

MSDS https://www.hit2lead.com/sds/4087770_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart