CH4415172753 ChemBridge Corporation

{[1-(4-morpholinylmethyl)cyclopropyl]methyl}amine dihydrochloride

  • MDL number MFCD27966314
  • Cas Number : 1559064-04-8
  • Molecular Weight : 243.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087899

Safety Information

MSDS https://www.hit2lead.com/sds/4087899_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart