CH4415172762 ChemBridge Corporation

2-cyclopropylalanine hydrate

  • MDL number MFCD27966323
  • Cas Number : 1820684-99-8
  • Molecular Weight : 147.1723

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087938

Safety Information

MSDS https://www.hit2lead.com/sds/4087938_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart