CH4415172764 ChemBridge Corporation

[(1-phenyl-1H-tetrazol-5-yl)methyl]amine hydrochloride hydrate

  • MDL number MFCD27966325
  • Molecular Weight : 229.6668

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087945

Safety Information

MSDS https://www.hit2lead.com/sds/4087945_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart