CH4415172768 ChemBridge Corporation

1-methyl-N-(4-pyridinylmethyl)-4-piperidinamine diethanedioate

  • MDL number MFCD27966329
  • Cas Number : 1559064-01-5
  • Molecular Weight : 385.3691

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2(COOH)2
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087955

Safety Information

MSDS https://www.hit2lead.com/sds/4087955_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart