CH4415172769 ChemBridge Corporation

N,N,N'-trimethyl-N'-4-piperidinyl-1,2-ethanediamine tri[(2Z)-2-butenedioate]

  • MDL number MFCD27966330
  • Cas Number : 1559122-13-2
  • Molecular Weight : 533.5262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3cis-HOOCCH=CHCOOH
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087956

Safety Information

MSDS https://www.hit2lead.com/sds/4087956_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart