CH4415172770 ChemBridge Corporation

N-methyl-1-(2-methyl-1,3-thiazol-4-yl)methanamine dihydrochloride hydrate

  • MDL number MFCD27966331
  • Molecular Weight : 233.1592

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087958

Safety Information

MSDS https://www.hit2lead.com/sds/4087958_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart