CH4415172771 ChemBridge Corporation

N-methyl-1-(8-quinolinyl)methanamine dihydrochloride dihydrate

  • MDL number MFCD27966332
  • Molecular Weight : 281.1788

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl 2H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087974

Safety Information

MSDS https://www.hit2lead.com/sds/4087974_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart