CH4415172774 ChemBridge Corporation

[(1-isopropyl-1,2,3,4-tetrahydro-6-quinolinyl)methyl]methylamine diethanedioate

  • MDL number MFCD27966335
  • Cas Number : 1559062-09-7
  • Molecular Weight : 398.4076

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2(COOH)2
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087980

Safety Information

MSDS https://www.hit2lead.com/sds/4087980_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart