CH4415172776 ChemBridge Corporation

1-(2-methoxybenzyl)piperazine bis(trifluoroacetate) hydrate

  • MDL number MFCD27966337
  • Molecular Weight : 452.3461

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2CF3COOH H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087982

Safety Information

MSDS https://www.hit2lead.com/sds/4087982_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart