CH4415172784 ChemBridge Corporation

N-(2-phenylethyl)-4-piperidinecarboxamide dihydrochloride

  • MDL number MFCD27966345
  • Cas Number : 1559059-74-3
  • Molecular Weight : 305.2433

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4088012

Safety Information

MSDS https://www.hit2lead.com/sds/4088012_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart