CH4415172785 ChemBridge Corporation

2-[4-(4-piperidinyl)-1-piperazinyl]ethanol hydrochloride

  • MDL number MFCD27966346
  • Cas Number : 1314088-18-0
  • Molecular Weight : 249.7808

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4088016

Safety Information

MSDS https://www.hit2lead.com/sds/4088016_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart