CH4415172792 ChemBridge Corporation

{2-[(4-methyl-4H-1,2,4-triazol-3-yl)thio]ethyl}amine hydrochloride

  • MDL number MFCD13193796
  • Cas Number : 1559064-02-6
  • Molecular Weight : 194.6856

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087757

Safety Information

MSDS https://www.hit2lead.com/sds/4087757_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart