CH4415172796 ChemBridge Corporation

[1-(4-methoxyphenyl)propyl]amine hydrochloride

  • MDL number MFCD06800956
  • Cas Number : 233608-13-4
  • Molecular Weight : 201.6931

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087761

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4087761
Availability details for
Quantity
  • Availability
  • Add to Cart