CH4415172801 ChemBridge Corporation

N,N-dimethyl-1-[4-(2-piperidinyl)phenyl]methanamine

  • MDL number MFCD09864210
  • Cas Number : 1177342-76-5
  • Molecular Weight : 218.3379

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087766

Safety Information

MSDS https://www.hit2lead.com/sds/4087766_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart