CH4415172809 ChemBridge Corporation

4-(benzyloxy)piperidine hydrochloride hydrate

  • MDL number MFCD13193814
  • Molecular Weight : 245.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087775

Safety Information

MSDS https://www.hit2lead.com/sds/4087775_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart