CH4415172811 ChemBridge Corporation

N-methyl-2-(4-methylphenoxy)ethanamine

  • MDL number MFCD06248189
  • Cas Number : 625437-29-8
  • Molecular Weight : 165.2322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087777

Safety Information

MSDS https://www.hit2lead.com/sds/4087777_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart