CH4415172814 ChemBridge Corporation

(8-methyl-5-quinolinyl)boronic acid

  • MDL number MFCD05256435
  • Cas Number : 1025010-58-5
  • Molecular Weight : 187.0029

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087780

Safety Information

MSDS https://www.hit2lead.com/sds/4087780_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart