CH4415172822 ChemBridge Corporation

2,7,8-trimethyl-4-quinolinecarboxylic acid

  • MDL number MFCD04035477
  • Cas Number : 436096-46-7
  • Molecular Weight : 215.2478

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087790

Safety Information

MSDS https://www.hit2lead.com/sds/4087790_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart