CH4415172825 ChemBridge Corporation

(1H-tetrazol-5-ylmethyl)amine hydrobromide

  • MDL number MFCD11983593
  • Cas Number : 33841-56-4
  • Molecular Weight : 180.0065

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087793

Safety Information

MSDS https://www.hit2lead.com/sds/4087793_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart