CH4415172827 ChemBridge Corporation

1-methyl-3-pyrrolidinecarboxylic acid hydrochloride

  • MDL number MFCD06738921
  • Cas Number : 50585-87-0
  • Molecular Weight : 165.618

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087795

Safety Information

MSDS https://www.hit2lead.com/sds/4087795_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart