CH4415172838 ChemBridge Corporation

[2-(1H-1,2,3-benzotriazol-1-yl)ethyl]amine hydrochloride

  • MDL number MFCD06739061
  • Cas Number : 2690-84-8
  • Molecular Weight : 198.6527

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087807

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4087807
Availability details for
Quantity
  • Availability
  • Add to Cart