CH4415172839 ChemBridge Corporation

N-methyl-3-(4-morpholinyl)-1-propanamine

  • MDL number MFCD09035683
  • Cas Number : 99114-72-4
  • Molecular Weight : 158.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087808

Safety Information

MSDS https://www.hit2lead.com/sds/4087808_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart