CH4415172844 ChemBridge Corporation

(4-methoxybenzyl)(2-methoxyethyl)amine hydrochloride

  • MDL number MFCD02026451
  • Cas Number : 1052593-01-7
  • Molecular Weight : 231.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087813

Safety Information

MSDS https://www.hit2lead.com/sds/4087813_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart