CH4415172845 ChemBridge Corporation

2-[(4-fluorobenzyl)amino]ethanol hydrochloride

  • MDL number MFCD07105351
  • Cas Number : 1093648-13-5
  • Molecular Weight : 205.657

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087814

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4087814
Availability details for
Quantity
  • Availability
  • Add to Cart