CH4415172847 ChemBridge Corporation

1-(2,3-dihydro-1H-inden-2-yl)-1,4-diazepane sulfate dihydrate

  • MDL number MFCD26959669
  • Molecular Weight : 350.431

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2SO4 2H2O
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087816

Safety Information

MSDS https://www.hit2lead.com/sds/4087816_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart