CH4415172851 ChemBridge Corporation

3-(4-piperidinyloxy)pyridine

  • MDL number MFCD08059774
  • Cas Number : 310881-48-2
  • Molecular Weight : 178.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087820

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4087820
Availability details for
Quantity
  • Availability
  • Add to Cart