CH4415172866 ChemBridge Corporation

[2-(dimethylamino)phenyl]boronic acid hydrochloride

  • MDL number MFCD07366476
  • Cas Number : 1315335-14-8
  • Molecular Weight : 201.4583

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087836

Safety Information

MSDS https://www.hit2lead.com/sds/4087836_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart