CH4415172875 ChemBridge Corporation

N-ethyl-2-(4-methyl-1-piperazinyl)ethanamine

  • MDL number MFCD08691507
  • Cas Number : 40172-12-1
  • Molecular Weight : 171.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087845

Safety Information

MSDS https://www.hit2lead.com/sds/4087845_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart