CH4415172876 ChemBridge Corporation

1-(6-chloro-1H-benzimidazol-2-yl)ethanone

  • MDL number MFCD08691510
  • Cas Number : 41510-16-1
  • Molecular Weight : 194.6177

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087846

Safety Information

MSDS https://www.hit2lead.com/sds/4087846_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart