CH4415172877 ChemBridge Corporation

3-(4,5,6,7-tetrahydro-2H-indazol-3-yl)propanoic acid

  • MDL number MFCD07643182
  • Cas Number : 196100-86-4
  • Molecular Weight : 194.2304

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087847

Safety Information

MSDS https://www.hit2lead.com/sds/4087847_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart