CH4415172886 ChemBridge Corporation

(1-isopropyl-4-piperidinyl)acetic acid hydrate

  • MDL number MFCD13031908
  • Cas Number : 1185436-47-8
  • Molecular Weight : 203.2786

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2O
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087856

Safety Information

MSDS https://www.hit2lead.com/sds/4087856_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart