CH4415172891 ChemBridge Corporation

[4-(4-morpholinyl)benzyl]amine dihydrochloride

  • MDL number MFCD11841266
  • Cas Number : 1185488-45-2
  • Molecular Weight : 265.1794

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087861

Safety Information

MSDS https://www.hit2lead.com/sds/4087861_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart