CH4415172892 ChemBridge Corporation

1-(aminocarbonyl)-3-piperidinecarboxylic acid

  • MDL number MFCD06799728
  • Cas Number : 871478-82-9
  • Molecular Weight : 172.1818

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4087862

Safety Information

MSDS https://www.hit2lead.com/sds/4087862_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart