CH4415172893 ChemBridge Corporation

1-(tert-butoxycarbonyl)-4-hydroxy-4-piperidinecarboxylic acid

  • MDL number MFCD08062531
  • Cas Number : 495414-64-7
  • Molecular Weight : 245.2723

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4045486

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4045486
Availability details for
Quantity
  • Availability
  • Add to Cart